Learn More
Medchemexpress LLC HY-17556 50mg , Folinic acid CAS:58-05-9 Purity:>98%
SDP
Catalog No. 502261006
Encompass_Preferred
HY-17556 50mg Folinic acid CAS: 58-05-9 Folinic acid (Leucovorin) is a biological folic acid and is generally administered along with Methotrexate (MTX) as a rescue agent to decrease MTX-induced toxicity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-1006 Supplier Medchemexpress LLC Supplier No. HY1755650MG
HY-17556 50mg Folinic acid CAS: 58-05-9 Folinic acid (Leucovorin) is a biological folic acid and is generally administered along with Methotrexate (MTX) as a rescue agent to decrease MTX-induced toxicity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.