Learn More
Medchemexpress LLC Folinic acid | 58-05-9 | MFCD00003666 | 100.0% | 473.44 | 100 MG
SDP
Catalog No. 5000386211
Encompass_Preferred
Folinic acid (Leucovorin) is a biological folic acid generally administered with Methotrexate as a rescue agent to reduce MTX-induced toxicity. It is for research use only.
  • Acts as a biological folic acid.
  • Administered as a rescue agent to decrease Methotrexate-induced toxicity.
  • Significantly reduces methotrexate-induced micronucleated binucleated cells and aberrant cells in vitro.
  • Appears to reverse growth inhibition following methotrexate administration in vivo.
Catalog No. 50-003-86211 Supplier Medchemexpress LLC Supplier No. HY17556100MG
Add to Cart
Add to Cart
Folinic acid (Leucovorin) is a biological folic acid generally administered with Methotrexate as a rescue agent to reduce MTX-induced toxicity. It is for research use only.
  • Acts as a biological folic acid.
  • Administered as a rescue agent to decrease Methotrexate-induced toxicity.
  • Significantly reduces methotrexate-induced micronucleated binucleated cells and aberrant cells in vitro.
  • Appears to reverse growth inhibition following methotrexate administration in vivo.