Learn More
Selleck Chemical LLC Folinic acid-5mg
SDP
Catalog No. 502389796
Encompass_Preferred
Folinic acid (Leucovorin) is a biological folic acid that increases the level of reduced folates in tissues, which promotes the inhibition of thymidylate synthase (TS).
Catalog No. 50-238-9796 Supplier Selleck Chemical LLC Supplier No. S57905MG
Folinic acid (Leucovorin) is a biological folic acid that increases the level of reduced folates in tissues, which promotes the inhibition of thymidylate synthase (TS).