Learn More
Medchemexpress LLC HY-17556 5mg Medchemexpress, Folinic acid CAS:58-05-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY175565MG
Encompass_Preferred
Medchemexpress, HY-17556 5mg Folinic acid CAS:58-05-9 Folinic acid (Leucovorin) is a biological folic acid and is generally administered along with methotrexate (MTX) as a rescue agent to decrease MTX-induced toxicity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1985