Learn More
Medchemexpress LLC HY-13664 100mg Medchemexpress, Folinic acid (calcium) CAS:1492-18-8 Purity:>98%
SDP
Catalog No. 502029019
Encompass_Preferred
Medchemexpress, HY-13664 100mg Folinic acid (calcium) CAS:1492-18-8 Folinic acid calcium (Leucovorin calcium) is a biological folic acid and is generally administered along with methotrexate (MTX) as a rescue agent to decrease MTX-induced toxicity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9019 Supplier Medchemexpress LLC Supplier No. HY13664100MG
Medchemexpress, HY-13664 100mg Folinic acid (calcium) CAS:1492-18-8 Folinic acid calcium (Leucovorin calcium) is a biological folic acid and is generally administered along with methotrexate (MTX) as a rescue agent to decrease MTX-induced toxicity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.