Learn More
Medchemexpress LLC Folinic acid calcium | 1492-18-8 | 99.86% | 50 MG
SDP
Catalog No. 5000348804
Encompass_Preferred
Folinic acid calcium (Leucovorin calcium) is a biological folic acid. It is commonly administered alongside methotrexate (MTX) as a rescue agent to mitigate MTX-induced toxicity.
  • For research use only.
  • Not sold to patients.
Catalog No. 50-003-48804 Supplier Medchemexpress LLC Supplier No. HY1366450MG
Add to Cart
Add to Cart
Folinic acid calcium (Leucovorin calcium) is a biological folic acid. It is commonly administered alongside methotrexate (MTX) as a rescue agent to mitigate MTX-induced toxicity.
  • For research use only.
  • Not sold to patients.