Learn More
Medchemexpress LLC Folinic acid (Standard) | 58-05-9 | 98.2% | 473.44 | 5 MG
SDP
Catalog No. 5000386486
Encompass_Preferred
Folinic acid (Standard) is the analytical standard of Folinic acid, intended for research and analytical applications. It is a biological folic acid administered with Methotrexate as a rescue agent to decrease Methotrexate-induced toxicity.
  • Intended for research and analytical applications.
  • Decreases Methotrexate-induced toxicity.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-003-86486 Supplier Medchemexpress LLC Supplier No. HY17556R5MG
Add to Cart
Add to Cart
Folinic acid (Standard) is the analytical standard of Folinic acid, intended for research and analytical applications. It is a biological folic acid administered with Methotrexate as a rescue agent to decrease Methotrexate-induced toxicity.
  • Intended for research and analytical applications.
  • Decreases Methotrexate-induced toxicity.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.