Learn More
Cayman Chemical Folipastatin, Purity: ≥95%, CAS: 139959-71-0, 2.5 mg
SDP
Catalog No. 502934141
Encompass_Preferred
An inhibitor of PLA2 ( IC50 = 39 μM); inhibits release of arachidonic acid from rat polymorphonuclear leukocytes (IC50 = 24 μM); inhibits radioligand binding to L-type calcium channels in porcine heart membranes (IC50 = 0.39 μM)
Catalog No. 50-293-4141 Supplier Cayman Chemical Supplier No. 249242.5 MG
An inhibitor of PLA2 ( IC50 = 39 μM); inhibits release of arachidonic acid from rat polymorphonuclear leukocytes (IC50 = 24 μM); inhibits radioligand binding to L-type calcium channels in porcine heart membranes (IC50 = 0.39 μM)