Learn More
Medchemexpress LLC HY-14769 1mg , Folitixorin CAS:3432-99-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY147691MG
Encompass_Preferred
HY-14769 1mg Folitixorin CAS: 3432-99-3 Folitixorin (5,10-methylenetetrahydrofolate) is a cofactor and an analog of leucovorin. Folitixorin is a promising agent for modulation of 5-FU cytotoxicity in adjuvant cancer research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1478