Learn More
Promocell GmbH PromoCell Follicle Dermal Papilla Cell Growth Medium 500 ml, ready-to-use, low-serum, with C-39625 attached to the flask
Catalog No. 50306257
Encompass
PromoCell Follicle Dermal Papilla Cell Growth Medium is a low-serum medium that has been developed for the in vitro cultivation of Human Follicle Dermal Papilla Cells.The medium is optimized for primary cultures of fetal, neonatal, and adult tissue, or for the maintenance of Follicle Dermal Papilla Cells.Components:* Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMix* Growth Medium Kit: Includes Basal Medium and SupplementPack* SupplementMix: Contains all media supplements premixed in one vial* SupplementPack: Contains all media supplements as individual vials| Media supplement | Final supplement concentration|| Fetal Calf Serum | 0.04 ml / ml || Bovine Pituitary Extract | 0.004 ml / ml || Basic Fibroblast Growth Factor (recombinant human) | 1 ng / ml || Insulin (recombinant human) | 5 μg / ml |
Catalog No. 50-306-257 Supplier Promocell GmbH Supplier No. C26501
PromoCell Follicle Dermal Papilla Cell Growth Medium is a low-serum medium that has been developed for the in vitro cultivation of Human Follicle Dermal Papilla Cells.The medium is optimized for primary cultures of fetal, neonatal, and adult tissue, or for the maintenance of Follicle Dermal Papilla Cells.Components:* Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMix* Growth Medium Kit: Includes Basal Medium and SupplementPack* SupplementMix: Contains all media supplements premixed in one vial* SupplementPack: Contains all media supplements as individual vials| Media supplement | Final supplement concentration|| Fetal Calf Serum | 0.04 ml / ml || Bovine Pituitary Extract | 0.004 ml / ml || Basic Fibroblast Growth Factor (recombinant human) | 1 ng / ml || Insulin (recombinant human) | 5 μg / ml |