Learn More
Creative Biomart Urofollitropin, Native Human Follicle-stimulating hormone (FSH)
Supplier:  Creative Biomart THP0145
Encompass_Preferred
The product is a purified form of follicle-stimulating hormone (FSH) that is manufactured by extraction from human urine and then purified. It consists of two non-covalently linked, non-identical glycoproteins designated as the alpha- and beta- subunits. The alpha- and beta- subunits have 92 and 111 amino acids. The alpha subunit is glycosylated at Asn 51 and Asn 78 while the beta subunit is glycosylated at Asn 7 and Asn 24. The product is important in the development of follicles produced by the ovaries.
Catalog No. 50-208-4196