Learn More
Creative Biomart Follitropin
Catalog No. 5000332697
Encompass
The product is a human follicle stimulating hormone (FSH) preparation of recombinant DNA origin which consists of two non-covalently linked non-identical glycoproteins designated as the alpha- and beta- subunits The alpha- and beta- subunits have 92 and 111 amino acids The alpha subunit is glycosylated at Asn 51 and Asn 78 while the beta subunit is glycosylated at Asn 7 and Asn 24 The product beta is produced in genetically engineered CHO
Catalog No. 50-003-32697 Supplier Creative Biomart Supplier No. THP0142
The product is a human follicle stimulating hormone (FSH) preparation of recombinant DNA origin which consists of two non-covalently linked non-identical glycoproteins designated as the alpha- and beta- subunits The alpha- and beta- subunits have 92 and 111 amino acids The alpha subunit is glycosylated at Asn 51 and Asn 78 while the beta subunit is glycosylated at Asn 7 and Asn 24 The product beta is produced in genetically engineered CHO