Learn More
EDVOTEK™ Follow That Flush Using Biotechnology to Stop Outbreaks
SDPSCI_ED
Catalog No. S000291 Shop All EDVOTEK Products
Change view
Click to view available options
Includes:
Biotech Tool
SYBR™
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S000291 SYBR™
S000290 Biotech Tool
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S000291 Supplier EDVOTEK™ Supplier No. 922S
Add to Cart
Add to Cart
Electrophoresis to test simulated water samples for bacteria and viruses
  • Students simulate real-world disease surveillance by analyzing wastewater for Norovirus and E. coli using gel electrophoresis
  • Hands-on use of core biotech tools to detect and identify pathogens in simulated water samples
  • Demonstrates how wastewater testing gives early warnings of outbreaks and informs public health responses
  • This project was made possible by a Science Education Partnership Award (SEPA),
  • Grant Number 2R44GM143977-02, from the National Institute of General Medical Sciences (NIGMS) of the National Institutes of Health (NIH)
  • Wastewater testing is a public health tool that helps scientists track specific diseases in a community
  • In this experiment, students use electrophoresis to test simulated water samples for bacteria and viruses
  • By learning how wastewater monitoring works, students see how biotechnology helps protect public health
Specifications
Certifications/Compliance NIH
Includes SYBR™
Class Size 8 Lab Groups
Product Type Electrophoresis Kit