Learn More
Aviva Systems Biology Corp FOLR1 Antibody - middle region (ARP41427_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41427_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against.LR1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FOLR1. Alternate names: FBP, FOLR, FR-alpha, MOv18ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57846