Learn More
ACROBiosystems ACROBiosystems Biotinylated Human FOLR1 Protein, Fc,Avitag™
Catalog No. 501957140
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human FOLR1, Fc,Avitag (FO1-H82F9) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (Accession # P15328-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 53.3 kDa. The protein migrates as 60-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Folic acid-BSA conjugate at 5 μg/mL (100 μL/well) can bind Biotinylated Human FOLR1, Fc,Avitag (Cat. No. FO1-H82F9) with a linear range of 0.02-0.313 μg/mL (QC tested).
Catalog No. 50-195-7140 Supplier ACROBiosystems Supplier No. FO1H82F925UG
Recombinant Protein;25UG;Biotinylated Human FOLR1, Fc,Avitag (FO1-H82F9) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (Accession # P15328-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 53.3 kDa. The protein migrates as 60-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Folic acid-BSA conjugate at 5 μg/mL (100 μL/well) can bind Biotinylated Human FOLR1, Fc,Avitag (Cat. No. FO1-H82F9) with a linear range of 0.02-0.313 μg/mL (QC tested).