Learn More
ACROBiosystems ACROBiosystems Human FOLR1 Protein, His Tag
Catalog No. 501956455
Encompass_Preferred
Recombinant Protein;1MG;Human FOLR1, His Tag (FO1-H52H1) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (Accession # P15328-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.5 kDa. The protein migrates as 35-40 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Folic Acid-BSA Conjugate at 5 μg/mL (100 μL/well) can bind Human FOLR1, His Tag (Cat. No. FO1-H52H1) with a linear range of 5-78 ng/mL (QC tested).
Catalog No. 50-195-6455 Supplier ACROBiosystems Supplier No. FO1H52H11MG
Recombinant Protein;1MG;Human FOLR1, His Tag (FO1-H52H1) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (Accession # P15328-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.5 kDa. The protein migrates as 35-40 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Folic Acid-BSA Conjugate at 5 μg/mL (100 μL/well) can bind Human FOLR1, His Tag (Cat. No. FO1-H52H1) with a linear range of 5-78 ng/mL (QC tested).