Learn More
Genscript Corporation FOLR1 His Cynomolgus/Rhesus
Catalog No. 5000056718
Encompass
Folate Receptor 1 (FOLR1) also known as Folate Receptor alpha and Folate Binding Protein (FBP) is a 37 - 42 kDa protein that mediates the cellular uptake of folic acid and reduced folates Dietary folates are required for many key metabolic processes including nucleotide and methionine synthesis the interconversion of glycine and serine and histidine breakdown FOLR1 binds to folate and reduced folic acid derivatives and mediates delivery of 5-methyltetrahydrofolate and folate analogs into the interior of cells Has high affinity for folate and folic acid analogs at neutral pH
Catalog No. 50-000-56718 Supplier Genscript Corporation Supplier No. Z03928100
Folate Receptor 1 (FOLR1) also known as Folate Receptor alpha and Folate Binding Protein (FBP) is a 37 - 42 kDa protein that mediates the cellular uptake of folic acid and reduced folates Dietary folates are required for many key metabolic processes including nucleotide and methionine synthesis the interconversion of glycine and serine and histidine breakdown FOLR1 binds to folate and reduced folic acid derivatives and mediates delivery of 5-methyltetrahydrofolate and folate analogs into the interior of cells Has high affinity for folate and folic acid analogs at neutral pH