Learn More
Sino Biological Cynomolgus / Rhesus FOLR1 Protein (His Tag) 50ug
Supplier:  Sino Biological 90950C08H50
Encompass_Preferred
A DNA sequence encoding the cynomolgus / rhesus FOLR1 (A0A2K5UR19/NP_001181576.1) (Met1-Ser234) was expressed with a polyhistidine tag at the C-terminus. Cynomolgus and Rhesus FOLR1 sequences are identical.
Catalog No. 50-211-5945