Learn More
ACROBiosystems Recombinant Protein;Human FOLR1 Protein, Fc Tag;HEK293;1MG;FO1,FOLR-1,FBP,FOLR,FOLR1
Catalog No. 501657761
Encompass_Preferred
Recombinant Protein;Human FOLR1 Protein, Fc Tag;1MG;Human FOLR1, Fc Tag (FO1-H5253) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (Accession # P15328-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.3 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Folic acid-BSA conjugate at 5 μg/mL (100 μL/well) can bind Human FOLR1, Fc Tag (Cat. No. FO1-H5253) with a linear range of 10-78 ng/mL (QC tested).
Catalog No. 50-165-7761 Supplier ACROBiosystems Supplier No. FO1H52531MG
Recombinant Protein;Human FOLR1 Protein, Fc Tag;1MG;Human FOLR1, Fc Tag (FO1-H5253) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (Accession # P15328-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.3 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Folic acid-BSA conjugate at 5 μg/mL (100 μL/well) can bind Human FOLR1, Fc Tag (Cat. No. FO1-H5253) with a linear range of 10-78 ng/mL (QC tested).