Learn More
ACROBiosystems Recombinant Protein;Rhesus macaque FOLR1 Protein, His tag;HEK293;100UG;FO1,FOLR-1,FBP,FOLR,FOLR1
Catalog No. 501658326
Encompass_Preferred
Recombinant Protein;Rhesus macaque FOLR1 Protein, His tag;100UG;Rhesus macaque FOLR1, His Tag (FO1-C52H8) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (Accession # NP_001181576.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.4 kDa. The protein migrates as 35-41 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Folic acid-BSA conjugate at 20 μg/mL (100 μL/well) can bind Rhesus macaque FOLR1, His Tag (Cat. No. FO1-C52H8) with a linear range of 0.156-2.5 μg/mL (QC tested).
Catalog No. 50-165-8326 Supplier ACROBiosystems Supplier No. FO1C52H8100UG
Recombinant Protein;Rhesus macaque FOLR1 Protein, His tag;100UG;Rhesus macaque FOLR1, His Tag (FO1-C52H8) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (Accession # NP_001181576.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.4 kDa. The protein migrates as 35-41 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Folic acid-BSA conjugate at 20 μg/mL (100 μL/well) can bind Rhesus macaque FOLR1, His Tag (Cat. No. FO1-C52H8) with a linear range of 0.156-2.5 μg/mL (QC tested).