Learn More
ACROBiosystems Recombinant Protein;Mouse FOLR1 Protein;HEK293;100UG;FO1,FOLR-1,FBP,FOLR,FOLR1
Catalog No. 501658413
Encompass_Preferred
Recombinant Protein;Mouse FOLR1 Protein;100UG;Mouse FOLR1, His Tag (FO1-M5225) is expressed from human 293 cells (HEK293). It contains AA Thr 25 - Ser 232 (Accession # P35846-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.2 kDa. The protein migrates as 35 kDa and 37 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Folic Acid-BSA Conjugate at 1 μg/mL (100 μL/well) can bind Mouse FOLR1, His Tag (Cat. No. FO1-M5225) with a linear range of 0.02-0.313 μg/mL (QC tested).
Catalog No. 50-165-8413 Supplier ACROBiosystems Supplier No. FO1M5225100UG
Recombinant Protein;Mouse FOLR1 Protein;100UG;Mouse FOLR1, His Tag (FO1-M5225) is expressed from human 293 cells (HEK293). It contains AA Thr 25 - Ser 232 (Accession # P35846-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.2 kDa. The protein migrates as 35 kDa and 37 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Folic Acid-BSA Conjugate at 1 μg/mL (100 μL/well) can bind Mouse FOLR1, His Tag (Cat. No. FO1-M5225) with a linear range of 0.02-0.313 μg/mL (QC tested).