Learn More
ACROBiosystems Recombinant Protein;Biotinylated Mouse FOLR1 Protein, Avi Tag (Avitag™);HEK293;200UG;FO1,FOLR-1,FBP,FOLR,FOLR1
Supplier:  ACROBiosystems FO1M82E9200UG
Encompass_Preferred
Recombinant Protein;Biotinylated Mouse FOLR1 Protein, Avi Tag (Avitag™);200UG;Biotinylated Mouse FOLR1, His Tag (FO1-M82E9) is expressed from human 293 cells (HEK293). It contains AA Thr 25 - Ser 232 (Accession # P35846-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 26.9 kDa. The protein migrates as 36 kDa and 44 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Folic acid-BSA at 5 μg/mL (100 μL/well) can bind Biotinylated Mouse FOLR1, His Tag (Cat. No. FO1-M82E9) with a linear range of 0.008-0.222 μg/mL (QC tested).
Catalog No. 50-165-8397