Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human FOLR1 Protein, His Tag, Avi Tag (Avitag™);HEK293;25UG;FO1,FOLR-1,FBP,FOLR,FOLR1
Supplier:  ACROBiosystems FO1H82E225UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human FOLR1 Protein, His Tag, Avi Tag (Avitag™);25UG;Biotinylated Human FOLR1, His Tag, Avi Tag (FO1-H82E2) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (Accession # P15328-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 28.3 kDa. The protein migrates as 38-42 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Folic acid-BSA conjugate at 5 μg/mL (100 μL/well) can bind Biotinylated Human FOLR1, His Tag, Avi Tag (Cat. No. FO1-H82E2) with a linear range of 8-125 ng/mL (QC tested).
Catalog No. 50-165-8007