Learn More
ACROBiosystems Recombinant Protein;Human FOLR2 Protein, His Tag;HEK293;100UG;FO2,FOLR2,FOLR-2,FOLR BETA,BETA-HFR,FBP/PL-1,FR-BETA,FR-P3,FBP
Supplier:  ACROBiosystems FO2H5223100UG
Encompass_Preferred
Recombinant Protein;Human FOLR2 Protein, His Tag;100UG;Human FOLR2, His Tag (FO2-H5223) is expressed from human 293 cells (HEK293). It contains AA Thr 17 - His 228 (Accession # P14207-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.5 kDa. The protein migrates as 30-35 kDa under reducing (R) condition (SDS-PAGE) due to N-linked glycosylation which has been verified by PNGase F digestion.;Immobilized Folic acid-BSA conjugate at 5 μg/mL (100 μL/well) can bind Human FOLR2, His Tag (Cat. No. FO2-H5223) with a linear range of 20-78 ng/mL (QC tested).
Catalog No. 50-165-7455