Learn More
Aviva Systems Biology Corp IZUMO1R Antibody - middle region (ARP59117_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59117_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against.LR4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human FOLR4. Alternate names: Folbp3ApplicationWB. Species reactivity: Guinea Pig, Horse, Human
Catalog No. 50-286-586