Learn More
Cole-Parmer Fomblin Y L-VAC 06/6 PFPE Pump Inert Oil, 1 kg
Supplier:  Cole-Parmer 7975170
Encompass_Preferred
The YL-Vac PFPE vacuum pump oils are formulated to ensure optimum pump performance and pump life They are ideal for use in chemical processes which require inert mechanical vacuum pump oils.
Catalog No. NC2061641