Learn More
Precision Plus Vacuum Parts Inc Fomblin AR 555 - 0.5oz Tube
Catalog No. 501956181
Encompass_Preferred
AR 555 is a high performance perfluorinated grease. AR 555 is deisgned for dry pump applications. Operating temperature range -20/250 degrees Celcius
Catalog No. 50-195-6181 Supplier Precision Plus Vacuum Parts Inc Supplier No. 870012
AR 555 is a high performance perfluorinated grease. AR 555 is deisgned for dry pump applications. Operating temperature range -20/250 degrees Celcius