Learn More
Kurt J Lesker Company Klübertemp GR RT 15N Vacuum Grease, 100G
SDP
Catalog No. NC0518800
Encompass
Klübertemp GR RT greases are used in numerous sectors of industry and a wide variety of components operating at high temperatures. Behaviour towards elastomers and plastics: Lubricants based on perfluorinated polyether oils and polytetrafluoroethylene are generally regarded as neutral towards most elastomers and plastics (exception: perfluorinated rubber). Nevertheless we recommend testing compatibility with the material to be used, especially prior to series application. Temp Range (-20 to +250 (°C))
Catalog No. NC0518800 Supplier Kurt J Lesker Company Supplier No. KLRT15F
Klübertemp GR RT greases are used in numerous sectors of industry and a wide variety of components operating at high temperatures. Behaviour towards elastomers and plastics: Lubricants based on perfluorinated polyether oils and polytetrafluoroethylene are generally regarded as neutral towards most elastomers and plastics (exception: perfluorinated rubber). Nevertheless we recommend testing compatibility with the material to be used, especially prior to series application. Temp Range (-20 to +250 (°C))