Learn More
Medchemexpress LLC Fomepizole | 7554-65-6 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0876500MG
Encompass_Preferred
Fomepizole (4-Methylpyrazole) is a potent cytochrome P450 (CYP2E1) inhibitor that acts as a competitive inhibitor of alcohol dehydrogenase. It blocks further conversion of methanol and ethylene glycol to toxic metabolites, making it a potential antidote for ethylene glycol or methanol poisoning.
  • Potent cytochrome P450 (CYP2E1) inhibitor
  • Competitive inhibitor of alcohol dehydrogenase
  • Blocks conversion of methanol and ethylene glycol to toxic metabolites
  • Potential antidote for ethylene glycol or methanol poisoning
Catalog No. 50-004-09156