Learn More
Medchemexpress LLC Fomepizole hydrochloride | 56010-88-9 | 99.9% | 1 ML
SDP
Catalog No. 5000407691
Encompass_Preferred
Fomepizole (4-Methylpyrazole) hydrochloride is a potent and orally active cytochrome P450 (CYP2E1) inhibitor. It is a competitive inhibitor of the enzyme alcohol dehydrogenase, blocking the conversion of methanol and ethylene glycol to toxic metabolites. It has the potential to be an antidote for ethylene glycol or methanol poisoning.
  • Potent and orally active cytochrome P450 (CYP2E1) inhibitor.
  • Competitive inhibitor of the enzyme alcohol dehydrogenase.
  • Blocks further conversion of methanol and ethylene glycol to toxic metabolites.
  • Potential for an antidote for ethylene glycol or methanol poisoning.
Catalog No. 50-004-07691 Supplier Medchemexpress LLC Supplier No. HYB0876A10MM/1ML
Fomepizole (4-Methylpyrazole) hydrochloride is a potent and orally active cytochrome P450 (CYP2E1) inhibitor. It is a competitive inhibitor of the enzyme alcohol dehydrogenase, blocking the conversion of methanol and ethylene glycol to toxic metabolites. It has the potential to be an antidote for ethylene glycol or methanol poisoning.
  • Potent and orally active cytochrome P450 (CYP2E1) inhibitor.
  • Competitive inhibitor of the enzyme alcohol dehydrogenase.
  • Blocks further conversion of methanol and ethylene glycol to toxic metabolites.
  • Potential for an antidote for ethylene glycol or methanol poisoning.