Learn More
Medchemexpress LLC Fomepizole (hydrochloride) | 56010-88-9 | 99.9% | 25 MG
SDP
Catalog No. 5000450812
Encompass_Preferred
Fomepizole hydrochloride (4-Methylpyrazole hydrochloride) is a potent and orally active cytochrome P450 (CYP2E1) inhibitor. It acts as a competitive inhibitor of the enzyme alcohol dehydrogenase, preventing the conversion of methanol and ethylene glycol into toxic metabolites. It is considered a potential antidote for ethylene glycol or methanol poisoning.
  • Potent and orally active cytochrome P450 (CYP2E1) inhibitor.
  • Competitive inhibitor of alcohol dehydrogenase.
  • Blocks conversion of methanol and ethylene glycol to toxic metabolites.
  • Potential antidote for ethylene glycol or methanol poisoning.
Catalog No. 50-004-50812 Supplier Medchemexpress LLC Supplier No. 25MGHYB0876A
Fomepizole hydrochloride (4-Methylpyrazole hydrochloride) is a potent and orally active cytochrome P450 (CYP2E1) inhibitor. It acts as a competitive inhibitor of the enzyme alcohol dehydrogenase, preventing the conversion of methanol and ethylene glycol into toxic metabolites. It is considered a potential antidote for ethylene glycol or methanol poisoning.
  • Potent and orally active cytochrome P450 (CYP2E1) inhibitor.
  • Competitive inhibitor of alcohol dehydrogenase.
  • Blocks conversion of methanol and ethylene glycol to toxic metabolites.
  • Potential antidote for ethylene glycol or methanol poisoning.