Learn More
Medchemexpress LLC Fomepizole hydrochloride | 56010-88-9 | MFCD00012707 | 99.9% | 100 MG
SDP
Catalog No. 5000408003
Encompass_Preferred
Fomepizole (4-Methylpyrazole) hydrochloride is a potent and orally active cytochrome P450 (CYP2E1) inhibitor. It is a competitive inhibitor of the enzyme alcohol dehydrogenase. Fomepizole hydrochloride blocks further conversion of methanol and ethylene glycol to toxic metabolites and has the potential for use as an antidote for ethylene glycol or methanol poisoning.
  • Potent and orally active cytochrome P450 (CYP2E1) inhibitor
  • Competitive inhibitor of alcohol dehydrogenase
  • Blocks conversion of methanol and ethylene glycol to toxic metabolites
  • Potential antidote for ethylene glycol or methanol poisoning
Catalog No. 50-004-08003 Supplier Medchemexpress LLC Supplier No. HYB0876A100MG
Fomepizole (4-Methylpyrazole) hydrochloride is a potent and orally active cytochrome P450 (CYP2E1) inhibitor. It is a competitive inhibitor of the enzyme alcohol dehydrogenase. Fomepizole hydrochloride blocks further conversion of methanol and ethylene glycol to toxic metabolites and has the potential for use as an antidote for ethylene glycol or methanol poisoning.
  • Potent and orally active cytochrome P450 (CYP2E1) inhibitor
  • Competitive inhibitor of alcohol dehydrogenase
  • Blocks conversion of methanol and ethylene glycol to toxic metabolites
  • Potential antidote for ethylene glycol or methanol poisoning