Learn More
Medchemexpress LLC HY-109528 10mg , Fomivirsen (sodium) CAS:160369-77-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10952810MG
Encompass_Preferred
HY-109528 10mg Fomivirsen (sodium) CAS: 160369-77-7 Fomivirsen sodium is an antisense 21 mer phosphorothioate oligonucleotide. Fomivirsen is an antiviral agent that is used cytomegalovirus retinitis (CMV) research, incluiding in AIDs. Fomivirsen sodium binds to and degrades the mRNAs encoding CMV immediate-early 2 protein (required for viral replication), thus providing bioactive effects for CMV retinitis by inhibition of virus proliferation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1285