Learn More
Cayman Chemical (E)-Anethol, Purity: ≥95%, CAS: 4180-23-8, 100 g
SDP
Catalog No. 502990231
Encompass_Preferred
A phenylpropanoid with antifungal and antioxidant activity; active against fermentatively growing S. cerevisiae under hypoxic, but not normoxic, conditions (MIC = 100 µg/ml), and against C. parapsilosis at 15% w/w; has antioxidant activity in a TEAC assay but does not scavenge DPPH radicals in a cell-free assay
Catalog No. 50-299-0231 Supplier Cayman Chemical Supplier No. 29862100 G
A phenylpropanoid with antifungal and antioxidant activity; active against fermentatively growing S. cerevisiae under hypoxic, but not normoxic, conditions (MIC = 100 µg/ml), and against C. parapsilosis at 15% w/w; has antioxidant activity in a TEAC assay but does not scavenge DPPH radicals in a cell-free assay