Learn More
TARGETMOL CHEMICALS INC FONDAPARINUX SODIUM 10MG
SDP
Catalog No. 502576815
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Fondaparinux sodium (SR-90107A) is an antithrombin-dependent factor Xa inhibitor with antithrombotic activity. purity: 98%
Catalog No. 50-257-6815 Supplier TARGETMOL CHEMICALS INC Supplier No. T407610MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Fondaparinux sodium (SR-90107A) is an antithrombin-dependent factor Xa inhibitor with antithrombotic activity. purity: 98%