Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fondaparinux sodium | 114870-03-0 | 98.0% | 1 ML
SDP

Catalog No. 5000408781
Encompass_Preferred

Fondaparinux sodium is an antithrombin-dependent factor Xa inhibitor intended for research use only. This compound exhibits high purity and is available as a white to off-white solid.

  • Antithrombin-dependent factor Xa inhibitor
  • For research use only
  • High purity of 98.0%
  • Appears as a white to off-white solid
  • Soluble in H2O
  • Exhibits a linear, dose-dependent pharmacokinetic profile
  • Features 100% bioavailability, rapid onset of action, and a half-life of 14 to 16 hours
  • Does not affect prothrombin time, activated partial thromboplastin time, platelet function, or aggregation

Catalog No. 50-004-08781 Supplier Medchemexpress LLC Supplier No. HYB059710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fondaparinux sodium is an antithrombin-dependent factor Xa inhibitor intended for research use only. This compound exhibits high purity and is available as a white to off-white solid.

  • Antithrombin-dependent factor Xa inhibitor
  • For research use only
  • High purity of 98.0%
  • Appears as a white to off-white solid
  • Soluble in H2O
  • Exhibits a linear, dose-dependent pharmacokinetic profile
  • Features 100% bioavailability, rapid onset of action, and a half-life of 14 to 16 hours
  • Does not affect prothrombin time, activated partial thromboplastin time, platelet function, or aggregation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.