Learn More
Medchemexpress LLC HY-B0597 50mg , Fondaparinux (sodium) Fondaparin sodium;SR-90107A CAS:114870-03-0 Purity:98%
SDP
Catalog No. 501493910
Encompass_Preferred
Medchemexpress, HY-B0597 50mg Fondaparinux (sodium) Fondaparin sodium;SR-90107A CAS:114870-03-0 Formula:C31H53N3Na10O49S8 Factor Xa Purity:98% Fondaparinux sodium is an antithrombin-dependent factor Xa inhibitor. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3910 Supplier Medchemexpress LLC Supplier No. HYB059750MG
Medchemexpress, HY-B0597 50mg Fondaparinux (sodium) Fondaparin sodium;SR-90107A CAS:114870-03-0 Formula:C31H53N3Na10O49S8 Factor Xa Purity:98% Fondaparinux sodium is an antithrombin-dependent factor Xa inhibitor. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.