Learn More
Crescent Chemical Co Inc FONOFOS 100 MG
SDP
Catalog No. 5073996
Encompass_Preferred
Fonofos Insecticide (WS) CAS 944-22-9 Molecular Weight - 246.3293 Storage - +5°C Catalog Num - DRE-C13900000 pesticide
Catalog No. 50-739-96 Supplier Crescent Chemical Co Inc Supplier No. PSC13900000
Fonofos Insecticide (WS) CAS 944-22-9 Molecular Weight - 246.3293 Storage - +5°C Catalog Num - DRE-C13900000 pesticide