Learn More
Diagnostic Biosystems FONTANA-MASSON Fontana-Masson
Catalog No. 502407525
Encompass_Preferred
The Fontana-Masson Stain Kit is intended for use in the histological visualization of Argentaffin cells and Melanin in paraffin or frozen sections.
Catalog No. 50-240-7525 Supplier Diagnostic Biosystems Supplier No. KT014
The Fontana-Masson Stain Kit is intended for use in the histological visualization of Argentaffin cells and Melanin in paraffin or frozen sections.