Learn More
Newcomer Supply Fontana Masson Stain Kit, 250 ml kit
SDP
Catalog No. NC2102259
Encompass
Newcomer Supply Fontana Masson Stain Kit procedure is used to demonstrate argentaffin substances such as melanin, argentaffin granules of carcinoid tumors, and some neurosecretory granules. This stain is not specific for melanin and argentaffin granules. Other reducing substances, such as formalin pigment, will also give a positive reaction.
Catalog No. NC2102259 Supplier Newcomer Supply Supplier No. 9105A
Newcomer Supply Fontana Masson Stain Kit procedure is used to demonstrate argentaffin substances such as melanin, argentaffin granules of carcinoid tumors, and some neurosecretory granules. This stain is not specific for melanin and argentaffin granules. Other reducing substances, such as formalin pigment, will also give a positive reaction.