Learn More
Cole-Parmer Food, Milk, and Dairy Tubing, 3/4" ID x 1" OD; 50 Ft
Supplier:  Cole-Parmer 5012173
Encompass_Preferred
Food, Milk, and Dairy Tubing is non-toxic, taste-free and odor-free, and meets all applicable regulatory standards for use in food, dairy and milk applications. Producers of food, milk and dairy products require the dependable performance of this tubing in countless filling, draining, transfer and processing applications. Smooth, non-porous and flexible, the Food, Milk, and Dairy tubing promotes a sanitary fluid path by maintaining an interior free of trapped particles, and minimizing potential for bacterial growth. In addition, it is resistant to harsh alkaline cleaners and is not affected by most commonly used sanitizers.
Catalog No. 50-256-3411