Learn More
MSC Delfin Industrial Hose, Anti-Static, Food Grade, 2 in, 10 ft
Catalog No. 502816531
Encompass
This Delfin Industrial hose is designed for industrial vacuum applications requiring specific safety and performance characteristics. It is anti-static, stretchable, and made from FDA food-grade materials to ensure safe and effective use in sensitive environments.
  • Anti-static properties
  • Stretchable design for flexibility
  • FDA food-grade compliant materials
  • Features a two in diameter
  • Offers a 10 ft length
  • White color
Catalog No. 50-281-6531 Supplier MSC Supplier No. 13676069
This Delfin Industrial hose is designed for industrial vacuum applications requiring specific safety and performance characteristics. It is anti-static, stretchable, and made from FDA food-grade materials to ensure safe and effective use in sensitive environments.
  • Anti-static properties
  • Stretchable design for flexibility
  • FDA food-grade compliant materials
  • Features a two in diameter
  • Offers a 10 ft length
  • White color