Learn More
MSC Delfin Food Grade Vacuum Hose, 50 mm, Conductive, 1 ea
Catalog No. 502818096
Encompass
Delfin Industrial FH0039 series conductive food grade vacuum hoses are engineered for rigorous cleaning in food processing environments. Designed to work with NRTL certified vacuum cleaners, these hoses are built from FDA acceptable materials, ensuring safety and compliance.
  • Food grade compliant with Class 2 Div II
  • Static dissipative polyurethane mitigates static electricity
  • Rigid red ABS helix maintains shape and prevents collapsing
  • Embedded copper grounding wire grounds the entire hose
  • Suitable for general cleaning of dust and conductive solids
  • Durable construction for long-lasting use
Catalog No. 50-281-8096 Supplier MSC Supplier No. 13676044
Delfin Industrial FH0039 series conductive food grade vacuum hoses are engineered for rigorous cleaning in food processing environments. Designed to work with NRTL certified vacuum cleaners, these hoses are built from FDA acceptable materials, ensuring safety and compliance.
  • Food grade compliant with Class 2 Div II
  • Static dissipative polyurethane mitigates static electricity
  • Rigid red ABS helix maintains shape and prevents collapsing
  • Embedded copper grounding wire grounds the entire hose
  • Suitable for general cleaning of dust and conductive solids
  • Durable construction for long-lasting use