Learn More
MSC Delfin Industrial Food Hose, 20 ft, 50 mm, 1 ea
Catalog No. 502818600
Encompass
Delfin Food Grade Vacuum Hose is a specially formulated static dissipative clear polyurethane hose designed for industrial vacuum applications in sensitive environments. It is reinforced with a rigid ABS black helix and an embedded copper grounding wire.
  • Black helix conductive food grade hose
  • Compliant with Class 2 Div. II standards
  • FDA acceptable materials
  • Includes hose cuffs
Catalog No. 50-281-8600 Supplier MSC Supplier No. 13676408
Delfin Food Grade Vacuum Hose is a specially formulated static dissipative clear polyurethane hose designed for industrial vacuum applications in sensitive environments. It is reinforced with a rigid ABS black helix and an embedded copper grounding wire.
  • Black helix conductive food grade hose
  • Compliant with Class 2 Div. II standards
  • FDA acceptable materials
  • Includes hose cuffs