Learn More
MSC Delfin Vacuum Hose, Conductive, Food Grade, 2 in, 1 ea
Catalog No. 502818972
Encompass
This vacuum hose is a conductive, food-grade solution for industrial cleaning applications. Constructed from static dissipative polyurethane with an embedded copper grounding wire, it is designed for use in environments requiring static control and food safety compliance.
  • Ideal for general cleaning of dust and conductive solids
  • Suitable for NRTL-certified vacuum cleaners in the food industry
  • Static dissipative polyurethane mitigates static electricity
  • Embedded copper wire grounds entire hose
  • Reinforced with a rigid black ABS helix to prevent collapsing
  • Made with FDA acceptable materials
  • Compliant with Class 2 Div II for hazardous environments
  • Hose cuffs included
Catalog No. 50-281-8972 Supplier MSC Supplier No. 13677471
This vacuum hose is a conductive, food-grade solution for industrial cleaning applications. Constructed from static dissipative polyurethane with an embedded copper grounding wire, it is designed for use in environments requiring static control and food safety compliance.
  • Ideal for general cleaning of dust and conductive solids
  • Suitable for NRTL-certified vacuum cleaners in the food industry
  • Static dissipative polyurethane mitigates static electricity
  • Embedded copper wire grounds entire hose
  • Reinforced with a rigid black ABS helix to prevent collapsing
  • Made with FDA acceptable materials
  • Compliant with Class 2 Div II for hazardous environments
  • Hose cuffs included