Learn More
MSC Delfin Conductive Food Grade, 50 mm, 20 ft, 1 ea
Catalog No. 502816991
Encompass
This conductive food grade hose is compliant with Class 2 Div II and ideal for general cleaning of dust and conductive solids, especially for NRTL-certified vacuum cleaners in the food industry. It is made from specially formulated static dissipative clear polyurethane with FDA acceptable materials.
  • Conductive food grade hose
  • Compliant with Class 2 Div II
  • Static dissipative clear polyurethane construction
  • Reinforced with a rigid green ABS helix
  • Embedded copper grounding wire
  • FDA acceptable materials
  • Includes hose cuffs
Catalog No. 50-281-6991 Supplier MSC Supplier No. 13676283
This conductive food grade hose is compliant with Class 2 Div II and ideal for general cleaning of dust and conductive solids, especially for NRTL-certified vacuum cleaners in the food industry. It is made from specially formulated static dissipative clear polyurethane with FDA acceptable materials.
  • Conductive food grade hose
  • Compliant with Class 2 Div II
  • Static dissipative clear polyurethane construction
  • Reinforced with a rigid green ABS helix
  • Embedded copper grounding wire
  • FDA acceptable materials
  • Includes hose cuffs