Learn More
MSC Delfin Industrial Vacuum Hose, Food Grade, Anti-Static, 1 ea
Catalog No. 502816992
Encompass
This vacuum hose features durable food-grade construction and static dissipative polyurethane to mitigate static electricity, ensuring safe use in food processing facilities. Its rigid ABS helix maintains shape and prevents collapsing, while an embedded copper wire grounds the entire hose.
  • Durable food-grade construction for use in food processing facilities
  • Static dissipative polyurethane mitigates static electricity and removes built-up charge
  • Rigid ABS helix maintains hose shape and prevents collapsing during operation
  • Embedded copper grounding wire for complete hose grounding
Catalog No. 50-281-6992 Supplier MSC Supplier No. 13676085
This vacuum hose features durable food-grade construction and static dissipative polyurethane to mitigate static electricity, ensuring safe use in food processing facilities. Its rigid ABS helix maintains shape and prevents collapsing, while an embedded copper wire grounds the entire hose.
  • Durable food-grade construction for use in food processing facilities
  • Static dissipative polyurethane mitigates static electricity and removes built-up charge
  • Rigid ABS helix maintains hose shape and prevents collapsing during operation
  • Embedded copper grounding wire for complete hose grounding