Learn More
METRIS INSTRUMENTS EAST LLC FOOD INSPECTOR FI40L FOR HACCP
Catalog No. 50320905
Encompass_Preferred
Thermometer, Infrared; Metric Instruments; For food inspection; HACCP; Range: -55 to 280 deg.C; 1.8F accuracy; Pre-set Emissivity; D:S: 12:1; Laser pointer; ~3 x 6 x 1.5 in.; 2 x AAA batteries; Battery life: 180 hrs.
Catalog No. 50-320-905 Supplier METRIS INSTRUMENTS EAST LLC Supplier No. FI40L
Thermometer, Infrared; Metric Instruments; For food inspection; HACCP; Range: -55 to 280 deg.C; 1.8F accuracy; Pre-set Emissivity; D:S: 12:1; Laser pointer; ~3 x 6 x 1.5 in.; 2 x AAA batteries; Battery life: 180 hrs.