Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PURELL™ Food Processing HEALTHY SOAP™ 0.5% PCMX Antimicrobial E2 Foam Handwash for CS6
SDP

Catalog No. 19044068 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Volume (Metric):
1200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
19-044-068 1200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-044-068 Supplier PURELL™ Supplier No. 658202
  • $113.00
    $85.65 / Case of 2

    Save $27.35

    24% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

One-step foam handwashing and sanitizing soap for the food processing industry.

  • Refill for PURELL™ CS6 Touch-Free Soap Dispensers
  • Removes fats, oils and other food processing soils
  • Certified Kosher
  • Dye-free and fragrance-free
  • SANITARY SEALED™ PET refill is easily recycled
  • SANITARY SEALED™ refill helps prevent contamination
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hand Cleanser
Color Clear
Form Foam
Size 1200 mL
Volume (Metric) 1200 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.