Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PURELL™ Food Processing HEALTHY SOAP™ 0.5% PCMX E2 Antimicrobial Foam
SDP

Catalog No. 19044064 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Volume (Metric):
1250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
19-044-064 1250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-044-064 Supplier PURELL™ Supplier No. 513204
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

One-step foam formula protects hands against common spoilage and foodborne illness organisms.

  • Ideal for food processing industry
  • Sanitary Sealed™ PET refill is easily recycled
  • Effectively removes fats, oils, and other food processing soils
  • Dye-free and fragrance-free
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hand Soap
Color Clear
Form Foam
Size 1250 mL
Volume (Metric) 1250 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.